Tisk…

Methyl-p-anisate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

253146-100G 253146-25G
SIAL253146-100GEA 2750 CZK
SIAL253146-100G SIAL253146-25G
Methyl-p-anisate, Sigma-Aldrich®
Methyl-p-anisate
Vzorec: C₉H₁₀O₃
MDL Number: MFCD00008437
CAS číslo: 121-98-2
EINECS: 204-513-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR