Tisk…

Ammonium trifluoroacetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

238732-25G 238732-100G
SIAL238732-25GEA 1880 CZK
SIAL238732-25G SIAL238732-100G
Ammonium trifluoroacetate, Sigma-Aldrich®
Ammonium trifluoroacetate
Vzorec: C₂H₄F₃NO₂
MDL Number: MFCD00012615
CAS číslo: 3336-58-1
EINECS: 222-072-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR