Butyl-akrylát, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
n-Butyl acrylate, Acrylic acid butyl ester
M-Clarity™ quality level = MQ200
234923-100ML
234923-18L
234923-1L
234923-2.5L
SIAL234923-100MLEA
1160
CZK
SIAL234923-100ML
SIAL234923-18L
SIAL234923-1L
SIAL234923-2.5L
Butyl-akrylát, Sigma-Aldrich®
Butyl-akrylát
Vzorec:
C₇H₁₂O₂ MW: 128,17 g/mol |
MDL Number:
MFCD00008809 CAS číslo: 141-32-2 EINECS: 205-480-7 UN: 2348 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...