Tisk…

Sodium dihydrogen citrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

234265-100G 234265-1KG
SIAL234265-100GEA 1170 CZK
SIAL234265-100G SIAL234265-1KG
Sodium dihydrogen citrate, Sigma-Aldrich®
Sodium dihydrogen citrate
Vzorec: C₆H₇NaO₇
Bod tání: 209…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00013067
CAS číslo: 18996-35-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR