Tisk…

Nitrosium tetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

175064-25G 175064-50G 175064-5G
SIAL175064-25GEA 8260 CZK
SIAL175064-25G SIAL175064-50G SIAL175064-5G
Nitrosium tetrafluoroborate, Sigma-Aldrich®
Nitrosium tetrafluoroborate
Vzorec: BF₄NO
MDL Number: MFCD00011433
CAS číslo: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR