3-Bromophenylhydrazinium chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride
M-Clarity™ quality level = MQ100
153958-25G
153958-5G
SIAL153958-25GEA
18620
CZK
SIAL153958-25G
SIAL153958-5G
3-Bromophenylhydrazinium chloride, Sigma-Aldrich®
3-Bromophenylhydrazinium chloride
Vzorec:
C₆H₈BrClN₂ MW: 223,5 g/mol |
MDL Number:
MFCD00012933 CAS číslo: 27246-81-7 EINECS: 248-362-0 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...