Tisk…

3,5-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride

M-Clarity™ quality level = MQ100

153648-10G 153648-50G
SIAL153648-10GEA 6550 CZK
SIAL153648-10G SIAL153648-50G
3,5-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®
3,5-Dichlorophenylhydrazinium chloride
Vzorec: C₆H₇Cl₃N₂
MW: 213,49 g/mol
MDL Number: MFCD00012938
CAS číslo: 63352-99-8
EINECS: 264-109-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR