Tisk…

2,6-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,6-Dichlorophenylhydrazine hydrochloride, 1-(2,6-Dichlorophenyl)hydrazine hydrochloride

M-Clarity™ quality level = MQ100

153419-10G 153419-1G
SIAL153419-10GEA 5740 CZK
SIAL153419-10G SIAL153419-1G
2,6-Dichlorophenylhydrazinium chloride, Sigma-Aldrich®
2,6-Dichlorophenylhydrazinium chloride
Vzorec: C₆H₇Cl₃N₂
MW: 213,49 g/mol
MDL Number: MFCD00012930
CAS číslo: 50709-36-9
EINECS: 256-730-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR