5-Methoxysalicylic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Hydroxy-5-methoxybenzoic acid
M-Clarity™ quality level = MQ100
146188-10G
146188-50G
SIAL146188-10GEA
3080
CZK
SIAL146188-10G
SIAL146188-50G
5-Methoxysalicylic acid, Sigma-Aldrich®
5-Methoxysalicylic acid
Vzorec:
C₈H₈O₄ MW: 168,15 g/mol Bod varu: 327 °C (1013 hPa) Bod tání: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002459 CAS číslo: 2612-02-4 EINECS: 220-037-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...