4-Bromophenylhydrazinium chloride, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
143219-10G
143219-50G
SIAL143219-10GEA
5390
CZK
SIAL143219-10G
SIAL143219-50G
4-Bromophenylhydrazinium chloride, Sigma-Aldrich®
4-Bromophenylhydrazinium chloride
Vzorec:
C₆H₈BrClN₂ |
MDL Number:
MFCD00012941 CAS číslo: 622-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...