2-Chloro-5-nitrobenzonitrile, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
135712-100G
135712-25G
SIAL135712-100GEA
9310
CZK
SIAL135712-100G
SIAL135712-25G
2-Chloro-5-nitrobenzonitrile, Sigma-Aldrich®
2-Chloro-5-nitrobenzonitrile
Vzorec:
C₇H₃ClN₂O₂ MW: 182,57 g/mol Bod varu: 119…122 °C (0,6 mmHg) Bod tání: 108 °C |
MDL Number:
MFCD00007292 CAS číslo: 16588-02-6 EINECS: 240-643-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...