Tisk…
Zinc(II) acetylacetonate hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Bis(2,4-pentanedionato-O,O')zinc (II) hydrate, Zinc acetylacetonate xhydrate, Zinc (II) 2,4-pentanedionate hydrate, Zinc (II) bis(2,4-pentanedionato-O,O') hydrate, Zinc acetylacetonate hydrate
M-Clarity™ quality level = MQ200
132306-100G 132306-500G
SIAL132306-100GEA 4080 CZK
SIAL132306-100G SIAL132306-500G
Zinc(II) acetylacetonate hydrate, Sigma-Aldrich®
Zinc(II) acetylacetonate hydrate
Vzorec: C₁₀H₁₄O₄Zn·H₂O
MDL Number: MFCD00149060
CAS číslo: 108503-47-5
EINECS: 237-860-3