Tisk…

2-Piperidinoethanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-(2-Hydroxyethyl)piperidine, 1-Piperidineethanol

M-Clarity™ quality level = MQ100

116068-100G 116068-500G
SIAL116068-100GEA 2690 CZK
SIAL116068-100G SIAL116068-500G
2-Piperidinoethanol, Sigma-Aldrich®
2-Piperidinoethanol
Vzorec: C₇H₁₅NO
MW: 129,2 g/mol
MDL Number: MFCD00006512
CAS číslo: 3040-44-6
EINECS: 221-244-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR