3-Bromo-4-fluorobenzonitrile ≥98%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A13318.06
A13318.14
A13318.06EA
862
CZK
A13318.06
A13318.14
3-Bromo-4-fluorobenzonitrile ≥98%
3-Bromo-4-fluorobenzonitrile
Purity: 98%
CAS No.: 79630-23-2
Molecular Formula: C7H3BrFN
Formula Weight: 200.01
Melting Point: 54-58°
Boiling Point: 134-136°/33mm
UN No.: 3439
MDL No.: MFCD00055432
Vzorec:
C₇H₃BrFN MW: 200,01 g/mol Bod varu: 134…136 °C (33 mmHg) Bod tání: 54…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055432 CAS číslo: 79630-23-2 EINECS: 279-200-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...