4-Bromo-2-fluorobenzaldehyde ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
A16112.06
A16112.14
A16112.22
A16112.06EA
1540
CZK
A16112.06
A16112.14
A16112.22
4-Bromo-2-fluorobenzaldehyde ≥97%
4-Bromo-2-fluorobenzaldehyde
Purity: 97%
CAS No.: 57848-46-1
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Melting Point: 59-63°
Boiling Point: 42°/19mm
MDL No.: MFCD00143261
Vzorec:
C₇H₄BrFO MW: 203,01 g/mol Bod varu: 42 °C (19 mmHg) Bod tání: 59…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143261 CAS číslo: 57848-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...