6-Chloro-m-anisidine ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
6-Chloro-3-anisidine, 2-Chloro-5-methoxyaniline, 3-Amino-4-chloroanisole
L20162.06
L20162.14
L20162.22
L20162.06EA
1830
CZK
L20162.06
L20162.14
L20162.22
6-Chloro-m-anisidine ≥98%
6-Chloro-m-anisidine
Purity: 98+%
CAS No.: 2401-24-3
Molecular Formula: C7H8ClNO
Formula Weight: 157.60
Refractive Index: 1.5865
Odour: Characteristic
Boiling Point: 135-137°/12mm
Beilstein Reference: 2082193
UN No.: 2233
MDL No.: MFCD00047830
Vzorec:
C₇H₈ClNO MW: 157.6 Bod varu: 135…137 °C (12 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00047830 CAS číslo: 2401-24-3 EINECS: 219-277-6 UN: 2233 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...