Tisk…

(S)-(+)-6-Methyl-5-hepten-2-ol ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

L18884.03 L18884.MD
L18884.03EA 5770 CZK
L18884.03 L18884.MD
(S)-(+)-6-Methyl-5-hepten-2-ol ≥99%
(S)-(+)-6-Methyl-5-hepten-2-ol

Purity: 99% CAS No.: 58917-26-3 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 67°(152°F) Refractive Index: 1.4480 Boiling Point: 78-80°/15mm Beilstein Reference: 1720071 Optical Rotation: +18° (neat) MDL No.: MFCD03093080

Vzorec: C₈H₁₆O
Bod varu: 78…80 °C (15 mmHg)
Hustota: 0,844
Bod vzplanutí: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD03093080
CAS číslo: 58917-26-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR