Tisk…
trans-1,4-Diphenyl-2-butene-1,4-dione predominantly trans 96%
Dodavatel: Thermo Fisher Scientific
Synonyms: 1,2-Dibenzoylethylene, trans-1,2-Dibenzoylethylene
L12643.03 L12643.06
L12643.03EA 759 CZK
L12643.03 L12643.06
trans-1,4-Diphenyl-2-butene-1,4-dione predominantly trans 96%
trans-1,4-Diphenyl-2-butene-1,4-dione
Purity: 96% CAS No.: 959-28-4 Molecular Formula: C16H12O2 Formula Weight: 236.27 Melting Point: 103-108° Beilstein Reference: 743958
Vzorec: C₁₆H₁₂O₂
MW: 236.27
Bod tání: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00003083
CAS číslo: 959-28-4
EINECS: 213-498-1