Sodium dihydrogen citrate ≥99%
Dodavatel: Thermo Scientific
Synonyms:
Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate
L12557.36
L12557.22
L12557.0P
L12557.36EA
1860
CZK
L12557.36
L12557.22
L12557.0P
Sodium dihydrogen citrate ≥99%
Sodium dihydrogen citrate
Purity: 99%
CAS No.: 18996-35-5
Molecular Formula: C6H7NaO7
Formula Weight: 214.11
Melting Point: 211-212°
Beilstein Reference: 3920246
Vzorec:
C₆H₇NaO₇ MW: 214,11 g/mol Bod tání: 211…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013067 CAS číslo: 18996-35-5 EINECS: 242-734-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...