Tisk…

(±)-6-Methyl-5-hepten-2-ol ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

L09181.06 L09181.14
L09181.06EA 1230 CZK
L09181.06 L09181.14
(±)-6-Methyl-5-hepten-2-ol ≥98%
(±)-6-Methyl-5-hepten-2-ol

Purity: 98% CAS No.: 1569-60-4 Molecular Formula: C8H16O Formula Weight: 128.22 Flash Point: 67°(152°F) Refractive Index: 1.4480 Boiling Point: 78-80°/15mm Beilstein Reference: 1720073 MDL No.: MFCD00004561

Vzorec: C₈H₁₆O
Bod varu: 78…80 °C (15 mmHg)
Hustota: 0,84
Bod vzplanutí: 67 °C (152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004561
CAS číslo: 1569-60-4
EINECS: 216-377-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR