Difenylsulfon ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Phenyl sulphone, sulphonyldibenzene
L08232.30
L08232.0B
L08232.30EA
1180
CZK
L08232.30
L08232.0B
Difenylsulfon ≥99%
Difenylsulfon
Purity: 99+%
CAS No.: 127-63-9
Molecular Formula: C12H10O2S
Formula Weight: 218.27
Flash Point: 184°(363°F)
Odour: Odourless
Boiling Point: 377-379°
Beilstein Reference: 1910573
Merck Reference: 14,3238
MDL No.: MFCD00007548
Vzorec:
C₁₂H₁₀O₂S MW: 218.28 Bod varu: 377…379 °C Bod tání: 123…130 °C Hustota: 1.36 Bod vzplanutí: 184 °C (363 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007548 CAS číslo: 127-63-9 EINECS: 204-853-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...