Tisk…

Bis(4-fluorophenyl)sulphone ≥98%

Dodavatel: Thermo Scientific

Synonyms: 4-Fluorophenyl sulphone, 4,4'-Difluoro diphenyl sulphone, 1-Fluor-4-((4-fluorphenyl)sulphonyl)benzol, 4,4'-sulphonylbis(fluorobenzene)

L04723.06 L04723.14
L04723.06EA 1080 CZK
L04723.06 L04723.14
Bis(4-fluorophenyl)sulphone ≥98%
Bis(4-fluorophenyl)sulphone

Purity: 98+% CAS No.: 383-29-9 Molecular Formula: C12H8F2O2S Formula Weight: 254.25 Odour: Odourless Melting Point: 97-101° Beilstein Reference: 1970911 MDL No.: MFCD00000350

Vzorec: C₁₂H₈F₂O₂S
MW: 254,26 g/mol
Bod tání: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00000350
CAS číslo: 383-29-9
EINECS: 206-847-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR