Butyl-akrylát ≥98% stabilizovaný
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
n-Butyl acrylate, Acrylic acid butyl ester
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
L03476.AE
L03476.AP
L03476.0F
L03476.AEEA
831
CZK
L03476.AE
L03476.AP
L03476.0F
Butyl-akrylát ≥98% stabilizovaný
Butyl-akrylát
Stabilized with up to 50 ppm 4-methoxyphenol
Purity: 98+%
CAS No.: 141-32-2
Molecular Formula: C7H12O2
Formula Weight: 128.17
Flash Point: 39°(102°F)
Refractive Index: 1.4180
Odour: Fruit-like
Melting Point: -64°
Boiling Point: 145-146°
Beilstein Reference: 1749970
Merck Reference: 14,1539
UN No.: 2348
MDL No.: MFCD00009446
Vzorec:
C₇H₁₂O₂ MW: 128,17 g/mol Bod varu: 145…146 °C Bod tání: –64 °C Hustota: 0,895 Bod vzplanutí: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008809 CAS číslo: 141-32-2 EINECS: 205-480-7 UN: 2348 ADR: 3,III Merck Index: 14,1539 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...