Tisk…

4-(Trifluoromethyl)phenylhydrazinium chloride 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: (4-(Trifluoromethyl)phenyl)hydrazine hydrochloride, 4-(Trifluoromethyl)phenylhydrazine hydrochloride, 4-Hydrazinobenzotrifluoride hydrochloride, 1-(4-(Trifluoromethyl)phenyl)hydrazine hydrochloride

H32340.03 H32340.09
H32340.03EA 1120 CZK
H32340.03 H32340.09
4-(Trifluoromethyl)phenylhydrazinium chloride 96%
4-(Trifluoromethyl)phenylhydrazinium chloride

Purity: 96% CAS No.: 2923-56-0 Molecular Formula: C7H7F3N2.HCl Formula Weight: 212.60 Melting Point: ca 210° dec. MDL No.: MFCD00204233

Vzorec: C₇H₈ClF₃N₂
MW: 212,6 g/mol
Bod tání: ∼ 210 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00204233
CAS číslo: 2923-56-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR