4-Methoxyanthranilic acid 96%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Amino-4-methoxybenzoic acid
H32319.03
H32319.06
H32319.03EA
1890
CZK
H32319.03
H32319.06
4-Methoxyanthranilic acid 96%
4-Methoxyanthranilic acid
Purity: 96%
CAS No.: 4294-95-5
Molecular Formula: C8H9NO3
Formula Weight: 167.16
Melting Point: ca 171° dec.
MDL No.: MFCD00667729
Vzorec:
C₈H₉NO₃ MW: 167,16 g/mol Bod tání: ∼ 171 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00667729 CAS číslo: 4294-95-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...