6-Bromo-2,3-difluorobenzaldehyde 98%
Dodavatel: Thermo Scientific
|
Varování
|
H31855.03
H31855.06
H31855.03EA
3990
CZK
H31855.03
H31855.06
6-Bromo-2,3-difluorobenzaldehyde 98%
6-Bromo-2,3-difluorobenzaldehyde
Purity: 98%
CAS No.: 360576-04-1
Molecular Formula: C7H3BrF2O
Formula Weight: 221.00
Melting Point: 39-43°
MDL No.: MFCD07368119
Vzorec:
C₇H₃BrF₂O Bod tání: 39…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368119 CAS číslo: 360576-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...