Tisk…
2,6-Difluoro-3-methoxyaniline ≥97%
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: 3-Amino-2,4-difluoroanisole
H26631.03 H26631.06
H26631.03EA 6150 CZK
H26631.03 H26631.06
2,6-Difluoro-3-methoxyaniline ≥97%
2,6-Difluoro-3-methoxyaniline
Purity: 97% CAS No.: 144851-62-7 Molecular Formula: C7H7F2NO Formula Weight: 159.14 Refractive Index: 1.5235
Vzorec: C₇H₇F₂NO
MW: 159.14
Storage Temperature: Ambient
MDL Number: MFCD04115898
CAS číslo: 144851-62-7
UN: 2810
ADR: 6.1 III