2-Amino-4-chlorothiophenol 96%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Amino-4-chlorobenzenethiol, 2-Amino-4-chlorophenyl mercaptan, 5-Chloro-2-mercaptoaniline
B25640.06
B25640.14
B25640.06EA
2380
CZK
B25640.06
B25640.14
2-Amino-4-chlorothiophenol 96%
2-Amino-4-chlorothiophenol
Purity: 96%
CAS No.: 1004-00-8
Molecular Formula: C6H6ClNS
Formula Weight: 159.64
Odour: Unpleasant
Melting Point: 52-59°
UN No.: 1759
MDL No.: MFCD00792528
Vzorec:
C₆H₆ClNS MW: 159,64 g/mol Bod tání: 52…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00792528 CAS číslo: 1004-00-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...