Tisk…
2,4-Dichloro-5-methoxyaniline ≥98%
Dodavatel: Thermo Fisher Scientific
Nebezpečí
Synonyms: 5-Amino-2,4-dichloroanisole, 4,6-Dichloro-m-anisidine
B25497.06 B25497.03 B25497.14
B25497.06EA 2020 CZK
B25497.06 B25497.03 B25497.14
2,4-Dichloro-5-methoxyaniline ≥98%
2,4-Dichloro-5-methoxyaniline
Purity: 98% CAS No.: 98446-49-2 Molecular Formula: C7H7Cl2NO Formula Weight: 192.05 UN No.: 2810 MDL No.: MFCD00974410
Vzorec: C₇H₇Cl₂NO
MW: 192.04
Storage Temperature: Ambient
MDL Number: MFCD00974410
CAS číslo: 98446-49-2
UN: 2811
ADR: 6.1 III