Tisk…

6-Methylpyridin-3-ol ≥99%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Methylpyridin-5-ol, 3-Hydroxy-6-methylpyridine, 5-Hydroxy-2-methylpyridine

B25014.06 B25014.14 B25014.22
B25014.06EA 779 CZK
B25014.06 B25014.14 B25014.22
6-Methylpyridin-3-ol ≥99%
6-Methylpyridin-3-ol

Purity: 99% CAS No.: 1121-78-4 Molecular Formula: C6H7NO Formula Weight: 109.13 Odour: Odourless Melting Point: 167-171° Beilstein Reference: 107077 MDL No.: MFCD00006339

Vzorec: C₆H₇NO
MW: 109,13 g/mol
Bod tání: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00006339
CAS číslo: 1121-78-4
EINECS: 214-337-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR