2-Methylpyridin-3-ol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Hydroxy-2-methylpyridine
B24944.03
B24944.06
B24944.03EA
841
CZK
B24944.03
B24944.06
2-Methylpyridin-3-ol ≥99%
2-Methylpyridin-3-ol
Purity: 99%
CAS No.: 1121-25-1
Molecular Formula: C6H7NO
Formula Weight: 109.13
Odour: Odourless
Melting Point: 166-169°
Beilstein Reference: 107937
MDL No.: MFCD00082538
Vzorec:
C₆H₇NO MW: 109.13 Bod tání: 166…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082538 CAS číslo: 1121-25-1 EINECS: 214-327-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...