Tisk…

2-Hydroxyethyl-akrylát ≥97% stabilizovaný

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Hydroxyethyl acrylate, Acrylic acid 2-hydroxyethyl ester

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)

B24815.0B B24815.30
B24815.0BEA 4570 CZK
B24815.0B B24815.30
2-Hydroxyethyl-akrylát ≥97% stabilizovaný
2-Hydroxyethyl-akrylát

Stabilized with 200-300 ppm 4-methoxyphenol


Purity: 97% CAS No.: 818-61-1 Molecular Formula: C5H8O3 Formula Weight: 116.12 Flash Point: 101°(214°F) Refractive Index: 1.4500 Odour: Pungent Melting Point: -60° Boiling Point: 192° UN No.: 2927 MDL No.: MFCD00002865

Vzorec: C₅H₈O₃
MW: 116,12 g/mol
Bod varu: 192 °C
Bod tání: < –60 °C
Hustota: 1,106
Bod vzplanutí: 101 °C (214 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002865
CAS číslo: 818-61-1
EINECS: 212-454-9
UN: 2922
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR