Tisk…

3-Bromophenylhydrazinium chloride ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Bromophenylhydrazine hydrochloride, 1-(3-Bromophenyl)hydrazine hydrochloride

B23510.06 B23510.14 B23510.22
B23510.06EA 2590 CZK
B23510.06 B23510.14 B23510.22
3-Bromophenylhydrazinium chloride ≥98%
3-Bromophenylhydrazinium chloride

Purity: 98% CAS No.: 27246-81-7 Molecular Formula: C6H7BrN2.HCl Formula Weight: 223.51 Melting Point: ca 225° dec. Beilstein Reference: 3565829

Vzorec: C₆H₈BrClN₂
MW: 223,5 g/mol
Bod tání: ∼ 225 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012933
CAS číslo: 27246-81-7
EINECS: 248-362-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR