Tisk…

Methyl-2-amino-4,5-dimethoxybenzoate 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Methyl 4,5-dimethoxyanthranilate, 2-Amino-4,5-dimethoxy benzoate, Methyl 2-amino-4,5-dimethoxybenzoate, 2-Amino-4,5-dimethoxybenzoic acid methyl ester

B21002.09 B21002.18
B21002.09EA 676 CZK
B21002.09 B21002.18
Methyl-2-amino-4,5-dimethoxybenzoate 98%
Methyl-2-amino-4,5-dimethoxybenzoate

Purity: 98% CAS No.: 26759-46-6 Molecular Formula: C10H13NO4 Formula Weight: 211.22 Melting Point: 128-130° Beilstein Reference: 2724754

Vzorec: C₁₀H₁₃NO₄
MW: 211,22 g/mol
Bod tání: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00014904
CAS číslo: 26759-46-6
EINECS: 247-967-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR