Benzyl cinnamate ≥99%
Dodavatel: Thermo Scientific
Synonyms:
Cinnamic acid benzyl ester, trans-Cinnamic acid benzyl ester, 3-Phenyl-2-propenoic acid benzyl ester
A19550.22
A19550.36
A19550.22EA
1140
CZK
A19550.22
A19550.36
Benzyl cinnamate ≥99%
Benzyl cinnamate
Purity: 99%
CAS No.: 103-41-3
Molecular Formula: C16H14O2
Formula Weight: 238.29
Flash Point: 180°(356°F)
Melting Point: 34-36°
Boiling Point: 195-200°/5mm
Beilstein Reference: 2051339
Merck Reference: 14,1130
MDL No.: MFCD00004789
Vzorec:
C₁₆H₁₄O₂ MW: 238,29 g/mol Bod varu: 195…200 °C (5 mmHg) Bod tání: 34…36 °C Hustota: 1,11 Bod vzplanutí: 180 °C (356 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004789 CAS číslo: 103-41-3 EINECS: 203-109-3 Merck Index: 14,1130 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...