Tisk…

Sodium toluene-4-sulphonate ≥90%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: p-Toluenesulphonic acid sodium salt, Sodium p-toluenesulphonate, Toluene-4-sulphonic acid sodium salt

A19332.18 A19332.30
A19332.18EA 1210 CZK
A19332.18 A19332.30
Sodium toluene-4-sulphonate ≥90%
Sodium toluene-4-sulphonate

Purity: 90+% CAS No.: 657-84-1 Molecular Formula: C7H7NaO3S Formula Weight: 194.19(anhy) Merck Reference: 14,9533 MDL No.: MFCD00798566

Vzorec: C₇H₇NaO₃S
MW: 194.19
Storage Temperature: Ambient
MDL Number: MFCD00798566
CAS číslo: 657-84-1
EINECS: 211-522-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR