2,2,2-Trifluoroethyl acrylate ≥98% stabilizovaný
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
Stabilized with 200 ppm 4-methoxyphenol
Purity: 98%
CAS No.: 407-47-6
Molecular Formula: C5H5F3O2
Formula Weight: 154.09
Storage Temperature: 2°C to 8°C
Flash Point: 12°(53°F)
Refractive Index: 1.3500
Odour: Pungent
Boiling Point: 91-93°
Beilstein Reference: 1756611
UN No.: 3272
MDL No.: MFCD00000444
Vzorec:
C₅H₅F₃O₂ Bod varu: 91…93 °C Hustota: 1,216 Bod vzplanutí: 12 °C (53 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000444 CAS číslo: 407-47-6 EINECS: 206-987-6 UN: 3272 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...