Ethyl (trimethylsilyl)acetate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ethyl 2-(trimethylsilyl)acetate, (Trimethylsilyl)acetic acid ethyl ester
A17707.09
A17707.18
A17707.30
A17707.09EA
1300
CZK
A17707.09
A17707.18
A17707.30
Ethyl (trimethylsilyl)acetate ≥98%
Ethyl (trimethylsilyl)acetate
Purity: 98+%
CAS No.: 4071-88-9
Molecular Formula: (CH3)3SiCH2CO2CH2CH3
Formula Weight: 160.29
Flash Point: 35°(95°F)
Refractive Index: 1.4150
Boiling Point: 156-158°
Beilstein Reference: 1755902
UN No.: 3272
MDL No.: MFCD00009172
Vzorec:
C₇H₁₆O₂Si MW: 160.29 Bod varu: 156…158 °C Hustota: 0.876 Bod vzplanutí: 35 °C (95 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009172 CAS číslo: 4071-88-9 EINECS: 223-783-2 UN: 3272 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...