Tisk…

Vinylacetát ≥99% stabilizovaný

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Acetic acid vinyl ester

Stabilization: Stabilised with hydroquinone (HQ)

A16247.0F A16247.AE A16247.AP
A16247.0FEA 2130 CZK
A16247.0F A16247.AE A16247.AP
Vinylacetát ≥99% stabilizovaný
Vinylacetát

Stabilized with 8-12 ppm hydroquinone


Purity: 99% CAS No.: 108-05-4 Molecular Formula: C4H6O2 Formula Weight: 86.09 Storage Temperature: 2°C to 8°C Flash Point: -8°(18°F) Physical Form: Clear liquid Density: 0.932 Refractive Index: 1.3960 Appearance: Colourless Odour: Sweet Melting Point: -100° Boiling Point: 72-73° Beilstein Reference: 1209327 Merck Reference: 14,9992 UN No.: 1301 MDL No.: MFCD00008713

Vzorec: C₄H₆O₂
MW: 86,09 g/mol
Bod varu: 72…73 °C
Bod tání: –100 °C
Hustota: 0,932
Bod vzplanutí: –8 °C (18 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008713
CAS číslo: 108-05-4
EINECS: 203-545-4
UN: 1301
ADR: 3,II
Merck Index: 14,9992

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR