4-Bromophenylhydrazinium chloride ≥97%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
(4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride
A15590.06
A15590.14
A15590.06EA
2320
CZK
A15590.06
A15590.14
4-Bromophenylhydrazinium chloride ≥97%
4-Bromophenylhydrazinium chloride
Purity: 97%
CAS No.: 622-88-8
Molecular Formula: C6H7BrN2.HCl
Formula Weight: 223.51
Odour: Odourless
Melting Point: ca 215° dec.
Beilstein Reference: 3565838
MDL No.: MFCD00012941
Vzorec:
C₆H₈BrClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD00012941 CAS číslo: 622-88-8 EINECS: 255-590-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...