Phenyl vinyl sulphone ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Vinyl phenyl sulphone, (Vinylsulphonyl)benzene
A14794.03
A14794.06
A14794.14
A14794.03EA
1090
CZK
A14794.03
A14794.06
A14794.14
Phenyl vinyl sulphone ≥99%
Phenyl vinyl sulphone
Purity: 99+%
CAS No.: 5535-48-8
Molecular Formula: C8H8O2S
Formula Weight: 168.22
Storage Temperature: 2°C to 8°C
Melting Point: 67-71°
Beilstein Reference: 1906894
MDL No.: MFCD00007554
Vzorec:
C₈H₈O₂S Bod tání: 67…71 °C |
MDL Number:
MFCD00007554 CAS číslo: 5535-48-8 EINECS: 226-890-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...