3-Chlorophenylhydrazinium chloride ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
1-(3-Chlorophenyl)hydrazine hydrochloride, 3-Chlorophenylhydrazine hydrochloride
A14770.09
A14770.18
A14770.30
A14770.09EA
1280
CZK
A14770.09
A14770.18
A14770.30
3-Chlorophenylhydrazinium chloride ≥97%
3-Chlorophenylhydrazinium chloride
Purity: 97%
CAS No.: 2312-23-4
Molecular Formula: C6H7ClN2.HCl
Formula Weight: 179.05
Odour: Odourless
Melting Point: ca 242° dec.
Beilstein Reference: 3565828
MDL No.: MFCD00012935
Vzorec:
C₆H₈Cl₂N₂ MW: 179,05 g/mol Bod tání: ∼ 242 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD02656655 CAS číslo: 2312-23-4 EINECS: 219-005-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...