Tisk…

p-Tolylhydrazinium chloride ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 4-Methylphenylhydrazinium chloride, 4-Tolylhydrazine hydrochloride, p-Tolylhydrazine hydrochloride, 1-(4-Methylphenyl)hydrazine hydrochloride, 4-Methylphenylhydrazine hydrochloride, 1-(4-Tolyl)hydrazine hydrochloride

A14220.06 A14220.14 A14220.22
A14220.06EA 1260 CZK
A14220.06 A14220.14 A14220.22
p-Tolylhydrazinium chloride ≥98%
p-Tolylhydrazinium chloride

Purity: 98% CAS No.: 637-60-5 Molecular Formula: C7H10N2.HCl Formula Weight: 158.63 Melting Point: ca 240° dec. Beilstein Reference: 3563996 UN No.: 2811 MDL No.: MFCD00012940

Vzorec: C₇H₁₁ClN₂
MW: 158,63 g/mol
Bod tání: ∼ 240 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00012940
CAS číslo: 637-60-5
EINECS: 211-295-2
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR