Disodium hydrogen citrate sesquihydrate ≥99%
Dodavatel: Thermo Fisher Scientific
Synonyms:
Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
A14001.30
A14001.0B
A14001.0I
A14001.30EA
944
CZK
A14001.30
A14001.0B
A14001.0I
Disodium hydrogen citrate sesquihydrate ≥99%
Disodium hydrogen citrate sesquihydrate
Purity: 99%
CAS No.: 6132-05-4
Molecular Formula: C6H6Na2O7.1.5H2O
Formula Weight: 263.11 (236.09anhy)
Melting Point: 149° dec.
MDL No.: MFCD00150445
Vzorec:
C₆H₆Na₂O₇·1.5H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150445 CAS číslo: 6132-05-4 EINECS: 205-623-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...