Methylakrylát ≥99% stabilised
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Acrylic acid methyl ester
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
Stabilized with ca 15 ppm 4-methoxyphenol
Purity: 99%
CAS No.: 96-33-3
Molecular Formula: CH2=CHCO2CH3
Formula Weight: 86.09
Storage Temperature: 2°C to 8°C
Flash Point: -3°(26°F)
Refractive Index: 1.4030
Odour: Stench
Melting Point: -75°
Boiling Point: 79-81°
Beilstein Reference: 605396
Merck Reference: 14,6011
UN No.: 1919
MDL No.: MFCD00008627
Vzorec:
C₄H₆O₂ MW: 86.09 Bod varu: 79…81 °C Bod tání: –75 °C Hustota: 0.956 Bod vzplanutí: –3 °C (26 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008627 CAS číslo: 96-33-3 EINECS: 202-500-6 UN: 1919 ADR: 3 II Merck Index: 14,6011 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...