Tisk…

Methyl-p-anisate ≥98%

Dodavatel: Thermo Fisher Scientific

A12598.14 A12598.22
A12598.14EA 1320 CZK
A12598.14 A12598.22
Methyl-p-anisate ≥98%
Methyl-p-anisate

Purity: 98+% CAS No.: 121-98-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Odour: Characteristic Boiling Point: 244-245° Beilstein Reference: 2208571 MDL No.: MFCD00008437

Vzorec: C₉H₁₀O₃
Bod varu: 244…245 °C
Bod tání: 48…51 °C
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008437
CAS číslo: 121-98-2
EINECS: 204-513-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR