Tisk…

Allyl trifluoroacetate ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A11690.09 A11690.18
A11690.09EA 1310 CZK
A11690.09 A11690.18
Allyl trifluoroacetate ≥97%
Allyl trifluoroacetate

Purity: 97% CAS No.: 383-67-5 Molecular Formula: CH2=CHCH2O2CCF3 Formula Weight: 154.09 Flash Point: -1°(30°F) Refractive Index: 1.3360 Boiling Point: 73-75° Beilstein Reference: 1766312 UN No.: 2924 MDL No.: MFCD00013567

Vzorec: C₅H₅F₃O₂
Bod varu: 73…75 °C
Hustota: 1,183
Bod vzplanutí: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013567
CAS číslo: 383-67-5
EINECS: 206-853-7
UN: 2924
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR