Tisk…

Oxamide ≥98%

Dodavatel: Thermo Scientific
Varování

A11570.0E A11570.22 A11570.36
A11570.0EEA 13870 CZK
A11570.0E A11570.22 A11570.36
Oxamide ≥98%
Oxamide

Purity: 98+% CAS No.: 471-46-5 Molecular Formula: C2H4N2O2 Formula Weight: 88.07 Odour: Odourless Melting Point: ca 350° dec. Beilstein Reference: 1743262 Merck Reference: 14,6918 MDL No.: MFCD00008007

Vzorec: C₂H₄N₂O₂
MW: 88,07 g/mol
Bod tání: ∼ 350 °C (rozkl.)
Hustota: 1,667
Storage Temperature: Ambient
MDL Number: MFCD00008007
CAS číslo: 471-46-5
EINECS: 207-442-5
Merck Index: 14,6918

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR