Allyl phenyl sulphone ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Phenyl 2-propenyl sulfone, Allylsulfonylbenzene, Allyl Phenyl sulphone, (Allylsulphonyl)benzene, (2-Propen-1-ylsulfonyl)benzene, (2-Propenylsulfonyl)benzene
A10401.06
A10401.14
A10401.06EA
1440
CZK
A10401.06
A10401.14
Allyl phenyl sulphone ≥98%
Allyl phenyl sulphone
Purity: 98%
CAS No.: 16212-05-8
Molecular Formula: C9H10O2S
Formula Weight: 182.24
Flash Point:
Vzorec:
C₉H₁₀O₂S MW: 182,24 g/mol Bod varu: 111…113 °C (0,5 mmHg) Hustota: 1,189 Bod vzplanutí: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014739 CAS číslo: 16212-05-8 EINECS: 240-338-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...