Tisk…

Silver trifluoroacetate ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: Silver(I) trifluoroacetate

13933.06 13933.14
13933.06EA 2120 CZK
13933.06 13933.14
Silver trifluoroacetate ≥98%
Silver trifluoroacetate

Purity: 98% CAS No.: 2966-50-9 Molecular Formula: C2AgF3O2 Formula Weight: 220.88 Physical Form: Powder Sensitivity: Light Sensitive and Hygroscopic Appearance: White to grey Odour: Odourless Melting Point: 257-260° dec. UN No.: 3077 MDL No.: MFCD00013199

Vzorec: C₂AgF₃O₂
MW: 220,88 g/mol
Bod tání: 257…260 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00013199
CAS číslo: 2966-50-9
EINECS: 221-004-0
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR